missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzonitrile 98.0+%, TCI America™

Supplier:  TCI America A1314250G

 View more versions of this product

Catalog No. A1314250G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Amino-5-nitrobenzonitrile
210°C
MFCD00007362
5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j
C1=CC(=C(C=C1[N+](=O)[O-])C#N)N
163.136
163.14
Crystalline Powder
17420-30-3
C7H5N3O2
250 g
MGCGMYPNXAFGFA-UHFFFAOYSA-N
2-amino-5-nitrobenzonitrile
28532
≥98.0% (GC)

Chemical Identifiers

17420-30-3
163.136
MGCGMYPNXAFGFA-UHFFFAOYSA-N
28532
C1=CC(=C(C=C1[N+](=O)[O-])C#N)N
C7H5N3O2
MFCD00007362
5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j
2-amino-5-nitrobenzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1806

RTECSNumber : BX2500000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions