missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzonitrile, 95%

Catalog No. p-7028224
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2541614 25 g
AAB2541622 100 g
AAB2541636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2541614 Supplier Thermo Scientific Chemicals Supplier No. B2541614
Only null left

CAS: 17420-30-3 | C7H5N3O2 | 163.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17420-30-3
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00007362
InChI Key MGCGMYPNXAFGFA-UHFFFAOYSA-N
Synonym 5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j
PubChem CID 28532
IUPAC Name 2-amino-5-nitrobenzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C#N)N

Specifications

Melting Point 207°C to 210°C
Quantity 25 g
Formula Weight 163.14
Percent Purity 95%
Chemical Name or Material 2-Amino-5-nitrobenzonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 241-446-8
RTECSNumber BX2500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only