missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzoic acid, 98%

Catalog No. AAA1042609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1042609 10 g
AAA1042618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1042609 Supplier Thermo Scientific Chemicals Supplier No. A1042609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 616-79-5 | C7H6N2O4 | 182.14 g/mol

2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to strong oxidizing agents. Stable under ordinary conditions.

Chemical Identifiers

CAS 616-79-5
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.14
MDL Number MFCD00017039
InChI Key RUCHWTKMOWXHLU-UHFFFAOYSA-N
Synonym 5-nitroanthranilic acid, benzoic acid, 2-amino-5-nitro, 2-amino-5-nitro-benzoic acid, 2-amino-5-nitrobenzoicacid, anthranilic acid, 5-nitro, 1-amino-2-carboxy-4-nitrobenzene, 2-amino-5-nitro-benzoicacid, pubchem14422, acmc-209mvd, 5-nitro anthranilic acid
PubChem CID 12032
ChEBI CHEBI:61280
IUPAC Name 2-amino-5-nitrobenzoic acid
SMILES NC1=CC=C(C=C1C(O)=O)[N+]([O-])=O

Specifications

Melting Point ∼270°C (decomposition)
Quantity 10 g
Beilstein 646219
Solubility Information Insoluble in water.
Formula Weight 182.14
Percent Purity 98%
Chemical Name or Material 2-Amino-5-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-493-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only