missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-n-propylsulfonylbenzimidazole, 98+%

Catalog No. p-7046259
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13711MC 100 mg
AAL1371103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13711MC Supplier Thermo Scientific Chemicals Supplier No. L13711MC
Only null left

CAS: 80983-34-2 | C10H13N3O2S | 239.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80983-34-2
Molecular Formula C10H13N3O2S
Molecular Weight (g/mol) 239.29
MDL Number MFCD01075656
InChI Key WTPBIYSMFKUQKY-UHFFFAOYSA-N
Synonym albendazole-2-aminosulfone, 2-amino-5-propylsulphonylbenzimidazole, 2-amino-5-n-propylsulfonylbenzimidazole, 1h-benzimidazol-2-amine, 5-propylsulfonyl, 5-propylsulfonyl-1h-benzo d imidazol-2-amine, 2-amino-5-n-propylsulphonylbenzimidazole, abz2nh2, 5-propylsulfonyl-1h-benzimidazol-2-amine, 5-propylsulphonyl-1h-benzimidazol-2-amine, 5-propane-1-sulfonyl-1h-1,3-benzodiazol-2-amine
PubChem CID 88125
ChEBI CHEBI:80621
IUPAC Name 6-propylsulfonyl-1H-benzimidazol-2-amine
SMILES CCCS(=O)(=O)C1=CC=C2N=C(N)NC2=C1

Specifications

Melting Point 220°C to 222°C
Quantity 100 mg
Formula Weight 239.3
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-n-propylsulfonylbenzimidazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only