CAS RN 80983-34-2
IUPAC Name:
6-(propane-1-sulfonyl)-1H-1,3-benzodiazol-2-amine
Synonyms:
5-(propane-1-sulfonyl)-3H-1,3-benzodiazol-2-amine
Molecular Weight (g/mol):
239.29
Molecular Formula:
C10H13N3O2S
InChi Key:
WTPBIYSMFKUQKY-UHFFFAOYSA-N
SMILES:
CCCS(=O)(=O)C1=CC=C2N=C(N)NC2=C1
1
–
1
of
1
results
1
Promotions Available
2-Amino-5-n-propylsulfonylbenzimidazole, 98+%
2-Amino-5-n-propylsulfonylbenzimidazole, 98+%
CAS: 80983-34-2 Molecular Formula: C10H13N3O2S Molecular Weight (g/mol): 239.29 MDL Number: MFCD01075656 InChI Key: WTPBIYSMFKUQKY-UHFFFAOYSA-N Synonym: albendazole-2-aminosulfone,2-amino-5-propylsulphonylbenzimidazole,2-amino-5-n-propylsulfonylbenzimidazole,1h-benzimidazol-2-amine, 5-propylsulfonyl,5-propylsulfonyl-1h-benzo d imidazol-2-amine,2-amino-5-n-propylsulphonylbenzimidazole,abz2nh2,5-propylsulfonyl-1h-benzimidazol-2-amine,5-propylsulphonyl-1h-benzimidazol-2-amine,5-propane-1-sulfonyl-1h-1,3-benzodiazol-2-amine PubChem CID: 88125 ChEBI: CHEBI:80621 IUPAC Name: 6-propylsulfonyl-1H-benzimidazol-2-amine SMILES: CCCS(=O)(=O)C1=CC=C2N=C(N)NC2=C1