missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylpyridine-3-carboxaldehyde, 95%

Catalog No. AAH66175MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66175MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH66175MD Supplier Thermo Scientific Chemicals Supplier No. H66175MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1023814-35-8 | C7H8N2O | 136.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1023814-35-8
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD09754122
InChI Key KHLBBDOMXLJWGR-UHFFFAOYSA-N
Synonym 2-amino-5-methylnicotinaldehyde, 2-amino-5-methylpyridine-3-carboxaldehyde
PubChem CID 32639818
IUPAC Name 2-amino-5-methylpyridine-3-carbaldehyde
SMILES CC1=CN=C(C(=C1)C=O)N

Specifications

Quantity 250 mg
Beilstein 23782189
Formula Weight 136.15
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-methylpyridine-3-carboxaldehyde
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only