CAS RN 1023814-35-8
IUPAC Name:
2-amino-5-methylpyridine-3-carbaldehyde
Synonyms:
2-amino-5-methylpyridine-3-carbaldehyde
Molecular Weight (g/mol):
136.15
Molecular Formula:
C7H8N2O
InChi Key:
KHLBBDOMXLJWGR-UHFFFAOYSA-N
SMILES:
CC1=CN=C(N)C(C=O)=C1
1
–
1
of
1
results
1
Promotions Available
2-Amino-5-methylpyridine-3-carboxaldehyde, 95%
2-Amino-5-methylpyridine-3-carboxaldehyde, 95%
CAS: 1023814-35-8 Molecular Formula: C7H8N2O Molecular Weight (g/mol): 136.154 MDL Number: MFCD09754122 InChI Key: KHLBBDOMXLJWGR-UHFFFAOYSA-N Synonym: 2-amino-5-methylnicotinaldehyde,2-amino-5-methylpyridine-3-carboxaldehyde PubChem CID: 32639818 IUPAC Name: 2-amino-5-methylpyridine-3-carbaldehyde SMILES: CC1=CN=C(C(=C1)C=O)N