missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylphenol, 98%

Catalog No. AAB2356009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2356009 10 g
AAB2356018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2356009 Supplier Thermo Scientific Chemicals Supplier No. B2356009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2835-98-5 | C7H9NO | 123.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2835-98-5
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007693
InChI Key HCPJEHJGFKWRFM-UHFFFAOYSA-N
Synonym 6-amino-m-cresol, 2-hydroxy-4-methylaniline, phenol, 2-amino-5-methyl, 4-amino-3-hydroxytoluene, 2-hydroxy-p-toluidine, unii-qcg4es2a26, 6-amino-3-cresol, 3-methyl-6-aminophenol, 6-amino-3-methylphenol, qcg4es2a26
PubChem CID 76082
IUPAC Name 2-amino-5-methylphenol
SMILES CC1=CC(=C(C=C1)N)O

Specifications

Melting Point 155°C to 159°C
Quantity 10 g
Beilstein 386144
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2-Amino-5-methylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-620-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only