missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7117424
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14215G 5 g
A142125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14215G Supplier TCI America Supplier No. A14215G
Only null left

Chemical Identifiers

CAS 2941-78-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007909
InChI Key NBUUUJWWOARGNW-UHFFFAOYSA-N
Synonym 5-methylanthranilic acid, 6-amino-m-toluic acid, 2-amino-5-methyl-benzoic acid, benzoic acid, 2-amino-5-methyl, m-toluic acid, 6-amino, 2-amino-5-methylbenzoicacid, unii-418xaj22v2, 2-amino-5-methyl benzoic acid, amino 2--5-methylbenzoic acid, pubchem4954
PubChem CID 76255
IUPAC Name 2-amino-5-methylbenzoic acid
SMILES CC1=CC=C(N)C(=C1)C(O)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-methylbenzoic Acid