missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylbenzoic acid, 97%

Catalog No. AAL0151306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0151306 5 g
AAL0151314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0151306 Supplier Thermo Scientific Chemicals Supplier No. L0151306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2941-78-8 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2941-78-8
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007909
InChI Key NBUUUJWWOARGNW-UHFFFAOYSA-N
Synonym 5-methylanthranilic acid, 6-amino-m-toluic acid, 2-amino-5-methyl-benzoic acid, benzoic acid, 2-amino-5-methyl, m-toluic acid, 6-amino, 2-amino-5-methylbenzoicacid, unii-418xaj22v2, 2-amino-5-methyl benzoic acid, amino 2--5-methylbenzoic acid, pubchem4954
PubChem CID 76255
IUPAC Name 2-amino-5-methylbenzoic acid
SMILES CC1=CC=C(N)C(=C1)C(O)=O

Specifications

Melting Point ∽175°C (decomposition)
Quantity 5 g
Beilstein 1101527
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 2-Amino-5-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-932-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only