missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylbenzenesulfonic acid, 99%

Catalog No. p-7027646
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2456030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2456030 Supplier Thermo Scientific Chemicals Supplier No. B2456030

CAS: 88-44-8 | C7H9NO3S | 187.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-44-8
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.21
MDL Number MFCD00007908
InChI Key LTPSRQRIPCVMKQ-UHFFFAOYSA-N
Synonym 4-aminotoluene-3-sulfonic acid, ptmsptmsa, ptms, ptmsa, benzenesulfonic acid, 2-amino-5-methyl, 6-amino-m-toluenesulfonic acid, red 4b acid, p-toluidine-2-sulfonic acid, p-toluidine-m-sulfonic acid, m-toluenesulfonic acid, 6-amino
PubChem CID 6934
IUPAC Name 2-amino-5-methylbenzenesulfonic acid
SMILES CC1=CC=C(N)C(=C1)S(O)(=O)=O

Specifications

Quantity 250 g
UN Number UN3261
Beilstein 2211509
Formula Weight 187.22
Percent Purity 99%
Chemical Name or Material 2-Amino-5-methylbenzenesulfonic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 201-831-3
RTECSNumber XT6320000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only