missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methyl-3-thiophenecarbonitrile 98.0+%, TCI America™

Catalog No. p-7117854
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23515G 5 g
A235125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23515G Supplier TCI America Supplier No. A23515G
Only null left

Chemical Identifiers

CAS 138564-58-6
Molecular Formula C6H6N2S
Molecular Weight (g/mol) 138.188
MDL Number MFCD01626055
InChI Key YGXADLPRHBRTPG-UHFFFAOYSA-N
Synonym 2-Amino-3-cyano-5-methylthiophene
PubChem CID 689056
IUPAC Name 2-amino-5-methylthiophene-3-carbonitrile
SMILES CC1=CC(=C(S1)N)C#N

Specifications

Melting Point 103°C
Quantity 5 g
Formula Weight 138.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-methyl-3-thiophenecarbonitrile