CAS RN 138564-58-6
IUPAC Name:
2-amino-5-methylthiophene-3-carbonitrile
Synonyms:
2-amino-5-methylthiophene-3-carbonitrile
Molecular Weight (g/mol):
138.19
Molecular Formula:
C6H6N2S
InChi Key:
YGXADLPRHBRTPG-UHFFFAOYSA-N
SMILES:
CC1=CC(C#N)=C(N)S1
1
–
1
of
1
results
1
2-Amino-5-methyl-3-thiophenecarbonitrile 98.0+%, TCI America™
CAS: 138564-58-6 Molecular Formula: C6H6N2S Molecular Weight (g/mol): 138.188 MDL Number: MFCD01626055 InChI Key: YGXADLPRHBRTPG-UHFFFAOYSA-N Synonym: 2-Amino-3-cyano-5-methylthiophene PubChem CID: 689056 IUPAC Name: 2-amino-5-methylthiophene-3-carbonitrile SMILES: CC1=CC(=C(S1)N)C#N