missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methoxypyrimidine, 97%

CAS: 13418-77-4 | C5H7N3O | 125.13 g/mol

Supplier:  Thermo Scientific Chemicals H6679903

 View more versions of this product

Catalog No. AAH6679903


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-5-methoxypyrimidine
White
MFCD00187864
607358
2-amino-5-methoxypyrimidine, 2-pyrimidinamine, 5-methoxy, 5-methoxy-2-pyrimidinamine, 2-pyrimidinamine, 5-methoxy-9ci, 5-methoxy-pyrimidin-2-ylamine, pubchem20801, enamine_000447, 2-pyrimidinamine,5-methoxy, 5-methoxypyrimidin-2-ylamine, ksc519a2h
COC1=CN=C(N)N=C1
125.13
125.13
Powder
13418-77-4
C5H7N3O
1 g
Air Sensitive
KAHHAPNRIQLSFT-UHFFFAOYSA-N
5-methoxypyrimidin-2-amine
2305244
97%

Chemical Identifiers

13418-77-4
125.13
KAHHAPNRIQLSFT-UHFFFAOYSA-N
5-methoxypyrimidin-2-amine
C5H7N3O
MFCD00187864
2305244
COC1=CN=C(N)N=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only