accessibility menu, dialog, popup

UserName

H3CONNNH2

CAS RN 13418-77-4

IUPAC Name: 5-methoxypyrimidin-2-amine
Synonyms: 5-methoxypyrimidin-2-amine
Molecular Weight (g/mol): 125.13
Molecular Formula: C5H7N3O
InChi Key: KAHHAPNRIQLSFT-UHFFFAOYSA-N
SMILES: COC1=CN=C(N)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results