missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methoxybenzoic acid, 98+%

Catalog No. p-7045331
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1029503 1 g
AAL1029506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1029503 Supplier Thermo Scientific Chemicals Supplier No. L1029503
Only null left

CAS: 6705-03-9 | C8H9NO3 | 167.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6705-03-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00016509
InChI Key UMKSAURFQFUULT-UHFFFAOYSA-N
Synonym 5-methoxyanthranilic acid, 2-amino-5-methoxybenzoicacid, 2-amino-5-methoxy-benzoic acid, benzoic acid, 2-amino-5-methoxy, pubchem2544, acmc-1b7jp, 5-methoxy anthranilic acid, 5-methoxyanththranillic acid, ksc495k4p, anthranilic acid, 5-methoxy
PubChem CID 277930
IUPAC Name 2-amino-5-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)N)C(=O)O

Specifications

Melting Point 150°C to 152°C
Quantity 1 g
Beilstein 777690
Formula Weight 167.16
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only