missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-mercapto-1,3,4-thiadiazole 98.0+%, TCI America™

Catalog No. A0425500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0425500G 500 g
A042525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0425500G Supplier TCI America Supplier No. A0425500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2349-67-9
Molecular Formula C2H3N3S2
Molecular Weight (g/mol) 133.187
MDL Number MFCD00003108
InChI Key GDGIVSREGUOIJZ-UHFFFAOYSA-N
Synonym 5-amino-1,3,4-thiadiazole-2-thiol, 2-amino-5-mercapto-1,3,4-thiadiazole, usaf pd-25, 1,3,4-thiadiazole-2 3h-thione, 5-amino, unii-b1heg7v21s, 5-amino-1,3,4-thiadiazole-2 3h-thione, 2-amino-1,3,4-thiadiazole-5-thiol, 2-thiol-5-amino-1,3,4-thiadiazole, ccris 6893, 2-mercapto-5-amino-1,3,4-thiadiazole
PubChem CID 2723847
IUPAC Name 5-amino-3H-1,3,4-thiadiazole-2-thione
SMILES C1(=NNC(=S)S1)N

Specifications

Melting Point 234°C
Color White-Yellow
Quantity 500 g
Formula Weight 133.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-mercapto-1,3,4-thiadiazole
EINECSNumber (5)-0570
RTECSNumber XI4550000
TSCA Yes