accessibility menu, dialog, popup

UserName

NH2NNHSS

CAS RN 2349-67-9

IUPAC Name: 5-amino-2,3-dihydro-1,3,4-thiadiazole-2-thione
Synonyms: 5-amino-3H-1,3,4-thiadiazole-2-thione
Molecular Weight (g/mol): 133.19
Molecular Formula: C2H3N3S2
InChi Key: GDGIVSREGUOIJZ-UHFFFAOYSA-N
SMILES: NC1=NNC(=S)S1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)