missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-mercapto-1,3,4-thiadiazole, 98%

Catalog No. p-7081460
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1145114 25 g
AAA1145122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1145114 Supplier Thermo Scientific Chemicals Supplier No. A1145114
Only null left

CAS: 2349-67-9 | C2H3N3S2 | 133.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2349-67-9
Molecular Formula C2H3N3S2
Molecular Weight (g/mol) 133.187
MDL Number MFCD00003108
InChI Key GDGIVSREGUOIJZ-UHFFFAOYSA-N
Synonym 5-amino-1,3,4-thiadiazole-2-thiol, 2-amino-5-mercapto-1,3,4-thiadiazole, usaf pd-25, 1,3,4-thiadiazole-2 3h-thione, 5-amino, unii-b1heg7v21s, 5-amino-1,3,4-thiadiazole-2 3h-thione, 2-amino-1,3,4-thiadiazole-5-thiol, 2-thiol-5-amino-1,3,4-thiadiazole, ccris 6893, 2-mercapto-5-amino-1,3,4-thiadiazole
PubChem CID 2723847
IUPAC Name 5-amino-3H-1,3,4-thiadiazole-2-thione
SMILES C1(=NNC(=S)S1)N

Specifications

Melting Point ∼243°C (decomposition)
Quantity 25 g
Beilstein 81728
Formula Weight 133.19
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-mercapto-1,3,4-thiadiazole
Hazard Category H315-H320-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-078-4
RTECSNumber XI4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only