missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodobenzoic acid, 98%

Catalog No. aaa1983814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1983814 25 g
AAA1983806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1983814 Supplier Thermo Scientific Chemicals Supplier No. A1983814
Only null left

CAS: 5326-47-6 | C7H6INO2 | 263.034 g/mol

2-Amino-5-iodobenzoic acid is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as pharmaceutical intermediates, polarizing films for Liquid Crystal Display (LCD) chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-iodobenzoic acid is used in wide range of medicals industrial applications as well as in human and animal nutrition products such as antiseptics and disinfectants, pharmaceutical intermediates, polarizing films for liquid crystal display [LCD] chemicals.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Light Sensitive. Store in the dark. Incompatible with light and oxidizing agents.

Chemical Identifiers

CAS 5326-47-6
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.034
MDL Number MFCD00007849
InChI Key GOLGILSVWFKZRQ-UHFFFAOYSA-N
Synonym 5-iodoanthranilic acid, benzoic acid, 2-amino-5-iodo, 5-iodoanthranil acid, anthranilic acid, 5-iodo, 2-amino-5-iodo-benzoic acid, pubchem2543, 5-iodo anthranilic acid, acmc-1ays8, 2-amino-5-iodobenzoicacid, 5-iodo-2-aminobenzoic acid
PubChem CID 72911
IUPAC Name 2-amino-5-iodobenzoic acid
SMILES C1=CC(=C(C=C1I)C(=O)O)N

Specifications

Melting Point ∼220°C (decomposition)
Quantity 25 g
Beilstein 639029
Sensitivity Light sensitive
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 263.03
Percent Purity 98%
Chemical Name or Material 2-Amino-5-iodobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-205-7
RTECSNumber DG2802000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only