missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodo-3-methylpyridine 98.0+%, TCI America™

Catalog No. A26835G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26835G 5 g
A26831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26835G Supplier TCI America Supplier No. A26835G

Chemical Identifiers

CAS 166266-19-9
Molecular Formula C6H7IN2
Molecular Weight (g/mol) 234.04
MDL Number MFCD02102422
InChI Key WTHKBDPHSGITFJ-UHFFFAOYSA-N
Synonym 2-amino-5-iodo-3-methylpyridine, 2-amino-5-iodo-3-picoline, 5-iodo-3-methyl-2-pyridinamine, 5-iodo-3-methyl-pyridin-2-ylamine, 2-amino-3-methyl-5-iodopyridine, 5-iodo-3-methyl-2-pyridylamine, 2-pyridinamine, 5-iodo-3-methyl, iodomethylpyridinamine, pubchem1264, acmc-209yui
PubChem CID 959586
IUPAC Name 5-iodo-3-methylpyridin-2-amine
SMILES CC1=CC(=CN=C1N)I

Specifications

Melting Point 111°C
Quantity 5 g
Formula Weight 234.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-iodo-3-methylpyridine