accessibility menu, dialog, popup

UserName

CH3IHNH2N

CAS RN 166266-19-9

IUPAC Name: 2-amino-5-iodo-3-methylpyridin-1-ium
Synonyms: 2-amino-5-iodo-3-methylpyridin-1-ium
Molecular Weight (g/mol): 235.05
Molecular Formula: C6H8IN2
InChi Key: WTHKBDPHSGITFJ-UHFFFAOYSA-O
SMILES: CC1=CC(I)=C[NH+]=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results