missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorophenol 97.0+%, TCI America™

Catalog No. A23001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23001G 1 g
A23005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23001G Supplier TCI America Supplier No. A23001G
Only null left

Chemical Identifiers

CAS 53981-24-1
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD00671759
InChI Key IIDUNAVOCYMUFB-UHFFFAOYSA-N
Synonym 4-fluoro-2-hydroxyaniline, phenol, 2-amino-5-fluoro, 2-amino-5-fluoro-phenol, 5-fluoro-2-aminophenol, 2-hydroxy-4-fluoroaniline, pubchem2828, acmc-1avob, 2-amino-5-flourophenol, 2-amino-5-fluor-phenol, 2-amino-5-fluoro phenol
PubChem CID 185763
IUPAC Name 2-amino-5-fluorophenol
SMILES C1=CC(=C(C=C1F)O)N

Specifications

Quantity 1 g
Formula Weight 127.12
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-fluorophenol