missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorobenzothiazole 98.0+%, TCI America™

Catalog No. A26821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26821G 1 g
A26825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26821G Supplier TCI America Supplier No. A26821G

Chemical Identifiers

CAS 20358-07-0
Molecular Formula C7H5FN2S
Molecular Weight (g/mol) 168.189
MDL Number MFCD05664560
InChI Key YHBIGBYIUMCLJS-UHFFFAOYSA-N
Synonym 2-amino-5-fluorobenzothiazole, 5-fluorobenzo d thiazol-2-amine, 2-benzothiazolamine, 5-fluoro, 5-fluoro-2-benzothiazolamine, 2-benzothiazolamine,5-fluoro, 5-fluorobenzothiazol-2-ylamine, pubchem21850, acmc-20a67y, 2-amino-5-fluorobenzothiozole, 5-fluoro-2-aminobenzothiazole
PubChem CID 821203
IUPAC Name 5-fluoro-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1F)N=C(S2)N

Specifications

Melting Point 177°C
Quantity 1 g
Formula Weight 168.19
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-fluorobenzothiazole