accessibility menu, dialog, popup

UserName

CAS RN 20358-07-0

IUPAC Name: 5-fluoro-1,3-benzothiazol-2-amine
Synonyms: 5-fluoro-1,3-benzothiazol-2-amine
Molecular Weight (g/mol): 168.19
Molecular Formula: C7H5FN2S
InChi Key: YHBIGBYIUMCLJS-UHFFFAOYSA-N
SMILES: NC1=NC2=CC(F)=CC=C2S1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)