missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluoro-4-methylpyridine, 95%

CAS: 301222-66-2 | C6H7FN2 | 126.134 g/mol

Supplier:  Thermo Scientific Chemicals H6481503

 View more versions of this product

Catalog No. AAH6481503


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-5-fluoro-4-methylpyridine
C6H7FN2
1 g
2-amino-5-fluoro-4-picoline, 2-amino-5-fluoro-4-methylpyridine, 5-fluoro-4-methyl-2-pyridinamine, 2-amino-4-methyl-5-fluoropyridine, pubchem1256, 5-fluoro-4-methyl-pyridin-2-amine, 2-pyridinamine,5-fluoro-4-methyl, 2-amino-4-methyl-5-fluoro pyridine, e 5-fluoro-4-methyl-2-pyridinamine, 2-pyridinamine, 5-fluoro-4-methyl-9ci
CC1=CC(=NC=C1F)N
126.134
126.13
301222-66-2
MFCD06659479
9694924
RKMOPXJNMQWPLH-UHFFFAOYSA-N
5-fluoro-4-methylpyridin-2-amine
22063670
95%

Chemical Identifiers

301222-66-2
126.134
RKMOPXJNMQWPLH-UHFFFAOYSA-N
22063670
CC1=CC(=NC=C1F)N
C6H7FN2
MFCD06659479
2-amino-5-fluoro-4-picoline, 2-amino-5-fluoro-4-methylpyridine, 5-fluoro-4-methyl-2-pyridinamine, 2-amino-4-methyl-5-fluoropyridine, pubchem1256, 5-fluoro-4-methyl-pyridin-2-amine, 2-pyridinamine,5-fluoro-4-methyl, 2-amino-4-methyl-5-fluoro pyridine, e 5-fluoro-4-methyl-2-pyridinamine, 2-pyridinamine, 5-fluoro-4-methyl-9ci
5-fluoro-4-methylpyridin-2-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only