missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-cyclopropyl-1,3,4-thiadiazole, 98%

Catalog No. AAL0577803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0577803 1 g
AAL0577806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0577803 Supplier Thermo Scientific Chemicals Supplier No. L0577803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57235-50-4 | C5H7N3S | 141.19 g/mol

2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.

Solubility
Soluble in Methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container. Store at -20°C.

Chemical Identifiers

CAS 57235-50-4
Molecular Formula C5H7N3S
Molecular Weight (g/mol) 141.19
MDL Number MFCD00051649
InChI Key AVLUMBXGKFNNAS-UHFFFAOYSA-N
Synonym 2-amino-5-cyclopropyl-1,3,4-thiadiazole, 5-cyclopropyl 1,3,4 thiadiazol-2-ylamine, 5-cyclopropyl-1,3,4-thiadiazole-2-ylamine, acmc-20ao4n, enamine_005503, 5-cyclopropyl-2-amino-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine,5-cyclopropyl, 5-cyclopropyl-1,3,4 thiadiazol-2-ylamine
PubChem CID 717021
IUPAC Name 5-cyclopropyl-1,3,4-thiadiazol-2-amine
SMILES NC1=NN=C(S1)C1CC1

Specifications

Melting Point 212°C to 216°C
Quantity 1 g
Beilstein 124356
Solubility Information Soluble in Methanol. Insoluble in water.
Formula Weight 141.2
Percent Purity 98%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2-Amino-5-cyclopropyl-1,3,4-thiadiazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only