accessibility menu, dialog, popup

UserName

NNSNH2

CAS RN 57235-50-4

IUPAC Name: 5-cyclopropyl-1,3,4-thiadiazol-2-amine
Synonyms: 5-cyclopropyl-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 141.19
Molecular Formula: C5H7N3S
InChi Key: AVLUMBXGKFNNAS-UHFFFAOYSA-N
SMILES: NC1=NN=C(S1)C1CC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)