missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-cyanopyridine 98.0+%, TCI America™

$200.21 - $800.87

Chemical Identifiers

CAS 4214-73-7
Molecular Formula C6H6N3
Molecular Weight (g/mol) 120.13
MDL Number MFCD00128902
InChI Key KDVBYUUGYXUXNL-UHFFFAOYSA-O
Synonym 2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
PubChem CID 818260
IUPAC Name 2-amino-5-cyanopyridin-1-ium
SMILES NC1=CC=C(C=[NH+]1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A16425G
View Documents
TCI America
A16425G
5 g
Each for $200.21
Only null left
 
A164225G
View Documents
TCI America
A164225G
25 g
Each for $800.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4214-73-7
120.13
KDVBYUUGYXUXNL-UHFFFAOYSA-O
818260
NC1=CC=C(C=[NH+]1)C#N
C6H6N3
MFCD00128902
2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
2-amino-5-cyanopyridin-1-ium

Specifications

4214-73-7
Yellow
C6H6N3
5 g
2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
NC1=CC=C(C=[NH+]1)C#N
120.13
119.13
Crystalline Powder
164°C
250°C
MFCD00128902
3439
KDVBYUUGYXUXNL-UHFFFAOYSA-O
2-amino-5-cyanopyridin-1-ium
818260
≥98.0% (GC,T)
2-Amino-5-cyanopyridine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents