missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-cyanopyridine, 98%

CAS: 4214-73-7 | C6H6N3 | 120.13 g/mol

$110.17 - $479.44

Chemical Identifiers

CAS 4214-73-7
Molecular Formula C6H6N3
Molecular Weight (g/mol) 120.13
MDL Number MFCD00128902
InChI Key KDVBYUUGYXUXNL-UHFFFAOYSA-O
Synonym 2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
PubChem CID 818260
IUPAC Name 6-aminopyridine-3-carbonitrile
SMILES NC1=CC=C(C=[NH+]1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC357620050
SDS View Documents
Thermo Scientific Chemicals
357620050
5 g Glass bottle
Each for $110.17
Only null left
 
AC357620250
SDS View Documents
Thermo Scientific Chemicals
357620250
25 g Glass bottle
Each for $479.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4214-73-7
120.13
KDVBYUUGYXUXNL-UHFFFAOYSA-O
818260
NC1=CC=C(C=[NH+]1)C#N
C6H6N3
MFCD00128902
2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
6-aminopyridine-3-carbonitrile

Specifications

4214-73-7
100.0
Brown
97.5% min. (GC)
C6H6N3
5 g
KDVBYUUGYXUXNL-UHFFFAOYSA-O
6-aminopyridine-3-carbonitrile
818260
98%
2-Amino-5-cyanopyridine, 98%
97.5
159°C to 165°C
Authentic
Glass bottle
MFCD00128902
2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
NC1=CC=C(C=[NH+]1)C#N
120.13
119.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only