missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-cyanopyridine, 98%

Catalog No. AAL1928206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1928206 5 g
AAL1928214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1928206 Supplier Thermo Scientific Chemicals Supplier No. L1928206

CAS: 4214-73-7 | C6H6N3 | 120.13 g/mol

2-Amino-5-cyanopyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-cyanopyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 4214-73-7
Molecular Formula C6H6N3
Molecular Weight (g/mol) 120.13
MDL Number MFCD00128902
InChI Key KDVBYUUGYXUXNL-UHFFFAOYSA-O
Synonym 2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
PubChem CID 818260
SMILES NC1=CC=C(C=[NH+]1)C#N

Specifications

Melting Point 159°C to 164°C
Boiling Point 240°C to 250°C (15 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 113876
Solubility Information Soluble in methanol. Insoluble in water.
IUPAC Name 2-amino-5-cyanopyridin-1-ium
Formula Weight 119.13
Percent Purity 98%
Chemical Name or Material 2-Amino-5-cyanopyridine
Hazard Category H301-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only