Learn More
2-Amino-5-chlorothiazole hydrochloride, 97%
CAS: 55506-37-1 | C3H4Cl2N2S | 171.04 g/mol
Supplier: Thermo Scientific Chemicals L0570706
Description
2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.
Solubility
Soluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications
| 2-Amino-5-chlorothiazole hydrochloride | |
| 168°C to 172°C | |
| MFCD00039006 | |
| 2-amino-5-chlorothiazole hydrochloride, 5-chlorothiazol-2-amine hydrochloride, 5-chloro-1,3-thiazol-2-amine hydrochloride, 2-thiazolamine, 5-chloro-, monohydrochloride, 5-chloro-2-thiazolamine hydrochloride, 2-amino-5-chloro-1,3-thiazole hydrochloride, 5-chlorothiazolamine hydrochloride, thiazole, 2-amino-5-chloro-hydrochloride, pubchem23694, c3h3cln2s.hcl | |
| GTMGFQYVLSQTKP-UHFFFAOYSA-N | |
| 5-chloro-1,3-thiazol-2-amine;hydrochloride | |
| 148743 | |
| 97% |
| 55506-37-1 | |
| C3H4Cl2N2S | |
| 5 g | |
| Soluble in water. | |
| [H+].[Cl-].NC1=NC=C(Cl)S1 | |
| 171.04 | |
| 171.05 |
Chemical Identifiers
| 55506-37-1 | |
| 171.04 | |
| GTMGFQYVLSQTKP-UHFFFAOYSA-N | |
| 148743 | |
| [H+].[Cl-].NC1=NC=C(Cl)S1 |
| C3H4Cl2N2S | |
| MFCD00039006 | |
| 2-amino-5-chlorothiazole hydrochloride, 5-chlorothiazol-2-amine hydrochloride, 5-chloro-1,3-thiazol-2-amine hydrochloride, 2-thiazolamine, 5-chloro-, monohydrochloride, 5-chloro-2-thiazolamine hydrochloride, 2-amino-5-chloro-1,3-thiazole hydrochloride, 5-chlorothiazolamine hydrochloride, thiazole, 2-amino-5-chloro-hydrochloride, pubchem23694, c3h3cln2s.hcl | |
| 5-chloro-1,3-thiazol-2-amine;hydrochloride |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 259-680-4
RTECSNumber : XJ1262800
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only