missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorothiazole hydrochloride, 97%

Catalog No. p-7044295
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0570703 1 g
AAL0570706 5 g
AAL0570714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0570703 Supplier Thermo Scientific Chemicals Supplier No. L0570703
Only null left

CAS: 55506-37-1 | C3H4Cl2N2S | 171.04 g/mol

2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.

Solubility
Soluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 55506-37-1
Molecular Formula C3H4Cl2N2S
Molecular Weight (g/mol) 171.04
MDL Number MFCD00039006
InChI Key GTMGFQYVLSQTKP-UHFFFAOYSA-N
Synonym 2-amino-5-chlorothiazole hydrochloride, 5-chlorothiazol-2-amine hydrochloride, 5-chloro-1,3-thiazol-2-amine hydrochloride, 2-thiazolamine, 5-chloro-, monohydrochloride, 5-chloro-2-thiazolamine hydrochloride, 2-amino-5-chloro-1,3-thiazole hydrochloride, 5-chlorothiazolamine hydrochloride, thiazole, 2-amino-5-chloro-hydrochloride, pubchem23694, c3h3cln2s.hcl
PubChem CID 148743
IUPAC Name 5-chloro-1,3-thiazol-2-amine;hydrochloride
SMILES [H+].[Cl-].NC1=NC=C(Cl)S1

Specifications

Melting Point 168°C to 172°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 171.05
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chlorothiazole hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 259-680-4
RTECSNumber XJ1262800
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only