missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloropyridine 98.0+%, TCI America™

Catalog No. p-7117081
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A083725G 25 g
A0837100G 100 g
A0837500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A083725G Supplier TCI America Supplier No. A083725G

Chemical Identifiers

CAS 1072-98-6
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00006324
InChI Key MAXBVGJEFDMHNV-UHFFFAOYSA-N
Synonym 2-amino-5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-2-aminopyridine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro, 2-amino-5-chloro pyridine, 2-amino5-chloropyridine
PubChem CID 66174
IUPAC Name 5-chloropyridin-2-amine
SMILES C1=CC(=NC=C1Cl)N

Specifications

Melting Point 136°C
Quantity 25 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloropyridine