missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloropyridine, 98%

Catalog No. p-7081886
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1206314 25 g
AAA1206322 100 g
AAA1206336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1206314 Supplier Thermo Scientific Chemicals Supplier No. A1206314
Only null left

CAS: 1072-98-6 | C5H5ClN2 | 128.559 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072-98-6
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00006324
InChI Key MAXBVGJEFDMHNV-UHFFFAOYSA-N
Synonym 2-amino-5-chloropyridine, 2-pyridinamine, 5-chloro, 5-chloro-2-pyridinamine, 5-chloro-2-pyridylamine, 5-chloro-2-aminopyridine, 5-chloro-pyridin-2-ylamine, amino 2--5-chloropyridine, pyridine, 2-amino-5-chloro, 2-amino-5-chloro pyridine, 2-amino5-chloropyridine
PubChem CID 66174
IUPAC Name 5-chloropyridin-2-amine
SMILES C1=CC(=NC=C1Cl)N

Specifications

Melting Point 135°C to 138°C
Boiling Point 127°C to 129°C (21 mmHg)
Flash Point 160°C (320°F)
Quantity 25 g
Beilstein 108735
Formula Weight 128.56
Percent Purity 98%
Chemical Name or Material 2-Amino-5-chloropyridine
Hazard Category H302
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 214-020-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only