missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloropyrazine 98.0+%, TCI America™

Catalog No. A27905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27905G 5 g
A27901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27905G Supplier TCI America Supplier No. A27905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33332-29-5
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD03423596
InChI Key HWCKAMAVEWVBDO-UHFFFAOYSA-N
Synonym 2-amino-5-chloropyrazine, pyrazinamine, 5-chloro, 5-amino-2-chloropyrazine, 5-chloro-pyrazin-2-ylamine, 2-amino-5-chloro-1,4-diazine, pubchem17501, acmc-1cix9, 5-chloro-2-amino-pyrazine, 5-chloro-2-pyrazinamine, 3-amino-6-chloropyrazine
PubChem CID 12808419
IUPAC Name 5-chloropyrazin-2-amine
SMILES C1=C(N=CC(=N1)Cl)N

Specifications

Melting Point 134°C
Color Yellow
Quantity 5 g
Formula Weight 129.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloropyrazine