accessibility menu, dialog, popup

UserName

NNClNH2

CAS RN 33332-29-5

IUPAC Name: 5-chloropyrazin-2-amine
Synonyms: pyrazinamine, 5-chloro-
Molecular Weight (g/mol): 129.55
Molecular Formula: C4H4ClN3
InChi Key: HWCKAMAVEWVBDO-UHFFFAOYSA-N
SMILES: NC1=CN=C(Cl)C=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)