missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloropyrazine, 95%

Catalog No. p-7057136
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6650906 5 g
AAH6650914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6650906 Supplier Thermo Scientific Chemicals Supplier No. H6650906

CAS: 33332-29-5 | C4H4ClN3 | 129.547 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33332-29-5
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.547
MDL Number MFCD03423596
InChI Key HWCKAMAVEWVBDO-UHFFFAOYSA-N
Synonym 2-amino-5-chloropyrazine, pyrazinamine, 5-chloro, 5-amino-2-chloropyrazine, 5-chloro-pyrazin-2-ylamine, 2-amino-5-chloro-1,4-diazine, pubchem17501, acmc-1cix9, 5-chloro-2-amino-pyrazine, 5-chloro-2-pyrazinamine, 3-amino-6-chloropyrazine
PubChem CID 12808419
IUPAC Name 5-chloropyrazin-2-amine
SMILES C1=C(N=CC(=N1)Cl)N

Specifications

Melting Point 122°C to 128°C
Quantity 5 g
Beilstein 507879
Formula Weight 129.55
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-chloropyrazine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only