missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloro-6-methylpyridine 98.0+%, TCI America™

Catalog No. A29201G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29201G 1 g
A29205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29201G Supplier TCI America Supplier No. A29201G

Chemical Identifiers

CAS 36936-23-9
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD09453335
InChI Key SHIKRPPKGCWKJO-UHFFFAOYSA-N
Synonym 2-amino-5-chloro-6-methylpyridine, 2-amino-5-chloro-6-picoline, 5-chloro-6-methyl-2-pyridinamine, 6-amino-3-chloropicoline, 2-amino-5-chloro-6-methyl pyridine, 5-chloro-6-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-chloro-6-methyl, pubchem5718, 6-amino-3-chloro-2-picoline, 5-chloro-6-methyl-pyridineamine
PubChem CID 12251007
IUPAC Name 5-chloro-6-methylpyridin-2-amine
SMILES CC1=C(C=CC(=N1)N)Cl

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 1 g
Formula Weight 142.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloro-6-methylpyridine