accessibility menu, dialog, popup

UserName

CH3NH2NCl

CAS RN 36936-23-9

IUPAC Name: 5-chloro-6-methylpyridin-2-amine
Synonyms: 5-chloro-6-methylpyridin-2-amine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: SHIKRPPKGCWKJO-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC(N)=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)