missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-chloro-3-nitropyridine 98.0+%, TCI America™
$72.70 - $218.10
Chemical Identifiers
| CAS | 5409-39-2 |
|---|---|
| Molecular Formula | C5H4ClN3O2 |
| Molecular Weight (g/mol) | 173.556 |
| MDL Number | MFCD00092011 |
| InChI Key | GILTXHIJUUIMPI-UHFFFAOYSA-N |
| Synonym | 2-amino-5-chloro-3-nitropyridine, 5-chloro-3-nitro-2-pyridinamine, 5-chloro-3-nitro-2 pyridylamine, 5-chloro-3-nitro-2-pyridylamine, 5-chloro-3-nitro-pyridin-2-amine, pubchem1151, acmc-1aqua, ksc496e3f, tpc-py065, 2-amino5-chloro-3-nitropyridine |
| PubChem CID | 224168 |
| IUPAC Name | 5-chloro-3-nitropyridin-2-amine |
| SMILES | C1=C(C(=NC=C1Cl)N)[N+](=O)[O-] |
Chemical Identifiers
| 5409-39-2 | |
| 173.556 | |
| GILTXHIJUUIMPI-UHFFFAOYSA-N | |
| 224168 | |
| C1=C(C(=NC=C1Cl)N)[N+](=O)[O-] |
| C5H4ClN3O2 | |
| MFCD00092011 | |
| 2-amino-5-chloro-3-nitropyridine, 5-chloro-3-nitro-2-pyridinamine, 5-chloro-3-nitro-2 pyridylamine, 5-chloro-3-nitro-2-pyridylamine, 5-chloro-3-nitro-pyridin-2-amine, pubchem1151, acmc-1aqua, ksc496e3f, tpc-py065, 2-amino5-chloro-3-nitropyridine | |
| 5-chloro-3-nitropyridin-2-amine |
Specifications
| 5409-39-2 | |
| C5H4ClN3O2 | |
| 5 g | |
| GILTXHIJUUIMPI-UHFFFAOYSA-N | |
| 5-chloro-3-nitropyridin-2-amine | |
| 224168 | |
| ≥98.0% (GC) | |
| 2-Amino-5-chloro-3-nitropyridine |
| 196°C | |
| MFCD00092011 | |
| 2-amino-5-chloro-3-nitropyridine, 5-chloro-3-nitro-2-pyridinamine, 5-chloro-3-nitro-2 pyridylamine, 5-chloro-3-nitro-2-pyridylamine, 5-chloro-3-nitro-pyridin-2-amine, pubchem1151, acmc-1aqua, ksc496e3f, tpc-py065, 2-amino5-chloro-3-nitropyridine | |
| C1=C(C(=NC=C1Cl)N)[N+](=O)[O-] | |
| 173.556 | |
| 173.56 | |
| Crystalline Powder |
Safety and Handling
TSCA : No