missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-chloro-3-methylbenzoic Acid 98.0+%, TCI America™
Supplier: TCI America A239925G
Specifications
| 2-Amino-5-chloro-3-methylbenzoic Acid | |
| 238°C | |
| MFCD02358895 | |
| 5-Chloro-3-methylanthranilic Acid, 2-Amino-5-chloro-m-toluic Acid | |
| CC1=C(C(=CC(=C1)Cl)C(=O)O)N | |
| 185.607 | |
| 185.61 | |
| Crystalline Powder |
| 20776-67-4 | |
| C8H8ClNO2 | |
| 25 g | |
| KOPXCQUAFDWYOE-UHFFFAOYSA-N | |
| 2-amino-5-chloro-3-methylbenzoic acid | |
| 11644044 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 20776-67-4 | |
| 185.607 | |
| KOPXCQUAFDWYOE-UHFFFAOYSA-N | |
| 11644044 | |
| CC1=C(C(=CC(=C1)Cl)C(=O)O)N |
| C8H8ClNO2 | |
| MFCD02358895 | |
| 5-Chloro-3-methylanthranilic Acid, 2-Amino-5-chloro-m-toluic Acid | |
| 2-amino-5-chloro-3-methylbenzoic acid |
Safety and Handling
TSCA : No