missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloro-3-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. A23995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23995G 5 g
A239925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23995G Supplier TCI America Supplier No. A23995G

Chemical Identifiers

CAS 20776-67-4
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD02358895
InChI Key KOPXCQUAFDWYOE-UHFFFAOYSA-N
Synonym 5-Chloro-3-methylanthranilic Acid, 2-Amino-5-chloro-m-toluic Acid
PubChem CID 11644044
IUPAC Name 2-amino-5-chloro-3-methylbenzoic acid
SMILES CC1=C(C(=CC(=C1)Cl)C(=O)O)N

Specifications

Melting Point 238°C
Quantity 5 g
Formula Weight 185.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chloro-3-methylbenzoic Acid