missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloro-3-fluoropyridine, 98%

Catalog No. AAH3013503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3013503 1 g
AAH3013506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3013503 Supplier Thermo Scientific Chemicals Supplier No. H3013503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 246847-98-3 | C5H4ClFN2 | 146.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 246847-98-3
Molecular Formula C5H4ClFN2
Molecular Weight (g/mol) 146.55
MDL Number MFCD09878434
InChI Key FPUPPVDVOFJSEP-UHFFFAOYSA-N
Synonym 2-amino-5-chloro-3-fluoropyridine, 5-chloro-3-fluoro-2-pyridinamine, 2-amino-3-fluoro-5-chloropyridine, 2-pyridinamine, 5-chloro-3-fluoro, 5-chloro-3-fluoro-2-pyridylamine, 5-chloro-3-fluoro-pyridin-2-amine, pubchem9633, acmc-1ceu9, ksc554c5t, 2-pyridinamine,5-chloro-3-fluoro
PubChem CID 11665433
IUPAC Name 5-chloro-3-fluoropyridin-2-amine
SMILES NC1=NC=C(Cl)C=C1F

Specifications

Melting Point 95°C to 97°C
Quantity 1 g
Formula Weight 146.55
Percent Purity 98%
Chemical Name or Material 2-Amino-5-chloro-3-fluoropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only