accessibility menu, dialog, popup

UserName

NClNH2F

CAS RN 246847-98-3

IUPAC Name: 5-chloro-3-fluoropyridin-2-amine
Synonyms: 5-chloro-3-fluoropyridin-2-amine
Molecular Weight (g/mol): 146.55
Molecular Formula: C5H4ClFN2
InChi Key: FPUPPVDVOFJSEP-UHFFFAOYSA-N
SMILES: NC1=NC=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results