missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromopyrazine 98.0+%, TCI America™

Catalog No. p-7117579
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16831G 1 g
A16835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16831G Supplier TCI America Supplier No. A16831G
Only null left

Chemical Identifiers

CAS 59489-71-3
Molecular Formula C4H4BrN3
Molecular Weight (g/mol) 174.001
InChI Key KRRTXVSBTPCDOS-UHFFFAOYSA-N
Synonym 2-amino-5-bromopyrazine, 5-bromo-2-pyrazinamine, 5-bromo-pyrazin-2-ylamine, 2-bromo-5-aminopyrazine, pyrazinamine, 5-bromo, 5-amino-2-bromopyrazine, 5-bromo-2-aminopyrazine, 5-bromopyrazin-2-ylamine, 5-bromopyrazinamine, 2-amino-5-bromo-pyrazine
PubChem CID 599539
IUPAC Name 5-bromopyrazin-2-amine
SMILES C1=C(N=CC(=N1)Br)N

Specifications

Melting Point 113°C
Quantity 1 g
Formula Weight 174.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromopyrazine