missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromobenzonitrile 98.0+%, TCI America™

Catalog No. p-7117658
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A19601G 1 g
A19605G 5 g
2 options

Catalog No. A19601G

Supplier: TCI America A19601G

Only null left

Chemical Identifiers

CAS 39263-32-6
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD00158946
InChI Key OATYCBHROMXWJO-UHFFFAOYSA-N
Synonym 4-bromo-2-cyanoaniline, 2-amino-5-brombenzonitrile, 2-amio-5-bromobenzonitrile, 5-bromoanthranilonitrile, 2-amino-5-bromobenzenecarbonitrile, 2-amino-5-bromo-benzonitrile, benzonitrile, 2-amino-5-bromo, 2-bromo-5-cyanoaniline, pubchem4602, 2-cyano-4-bromoaniline
PubChem CID 429740
IUPAC Name 2-amino-5-bromobenzonitrile
SMILES NC1=CC=C(Br)C=C1C#N

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 197.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromobenzonitrile