Learn More
2-Amino-5-bromobenzonitrile, 96%
CAS: 39263-32-6 | C7H5BrN2 | 197.04 g/mol
$90.56 - $312.47
Chemical Identifiers
| CAS | 39263-32-6 |
|---|---|
| Molecular Formula | C7H5BrN2 |
| Molecular Weight (g/mol) | 197.04 |
| MDL Number | MFCD00158946 |
| InChI Key | OATYCBHROMXWJO-UHFFFAOYSA-N |
| Synonym | 4-bromo-2-cyanoaniline, 2-amino-5-brombenzonitrile, 2-amio-5-bromobenzonitrile, 5-bromoanthranilonitrile, 2-amino-5-bromobenzenecarbonitrile, 2-amino-5-bromo-benzonitrile, benzonitrile, 2-amino-5-bromo, 2-bromo-5-cyanoaniline, pubchem4602, 2-cyano-4-bromoaniline |
| PubChem CID | 429740 |
| IUPAC Name | 2-amino-5-bromobenzonitrile |
| SMILES | NC1=CC=C(Br)C=C1C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5521703
|
Thermo Scientific Chemicals
H5521703 |
1 g |
Each for $90.56
|
|
|||||
|
AAH5521706
|
Thermo Scientific Chemicals
H5521706 |
5 g |
Each for $312.47
|
|
|||||
Description
2-Amino-5-bromobenzonitrile can react with Oxalic acid dimethyl ester to get 5-Bromo-N-methylanthranilonitrile.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-5-bromobenzonitrile can react with Oxalic acid dimethyl ester to get 5-Bromo-N-methylanthranilonitrile.
Solubility
Soluble in methanol.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
| 39263-32-6 | |
| 197.04 | |
| OATYCBHROMXWJO-UHFFFAOYSA-N | |
| 429740 | |
| NC1=CC=C(Br)C=C1C#N |
| C7H5BrN2 | |
| MFCD00158946 | |
| 4-bromo-2-cyanoaniline, 2-amino-5-brombenzonitrile, 2-amio-5-bromobenzonitrile, 5-bromoanthranilonitrile, 2-amino-5-bromobenzenecarbonitrile, 2-amino-5-bromo-benzonitrile, benzonitrile, 2-amino-5-bromo, 2-bromo-5-cyanoaniline, pubchem4602, 2-cyano-4-bromoaniline | |
| 2-amino-5-bromobenzonitrile |
Specifications
| 39263-32-6 | |
| C7H5BrN2 | |
| 1 g | |
| 4-bromo-2-cyanoaniline, 2-amino-5-brombenzonitrile, 2-amio-5-bromobenzonitrile, 5-bromoanthranilonitrile, 2-amino-5-bromobenzenecarbonitrile, 2-amino-5-bromo-benzonitrile, benzonitrile, 2-amino-5-bromo, 2-bromo-5-cyanoaniline, pubchem4602, 2-cyano-4-bromoaniline | |
| OATYCBHROMXWJO-UHFFFAOYSA-N | |
| 2-amino-5-bromobenzonitrile | |
| 429740 | |
| 96% |
| 96°C to 100°C | |
| MFCD00158946 | |
| UN3439 | |
| Soluble in methanol. | |
| NC1=CC=C(Br)C=C1C#N | |
| 197.04 | |
| 197.03 | |
| 2-Amino-5-bromobenzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 254-387-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only