missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromobenzoic acid, 98%

Catalog No. p-7081772
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1191506 5 g
AAA1191514 25 g
AAA1191522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1191506 Supplier Thermo Scientific Chemicals Supplier No. A1191506

CAS: 5794-88-7 | C7H6BrNO2 | 216.034 g/mol

It is used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. Also used in the determination of cobalt, copper, nickel, and zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of hepatitis C virus NS5b RNA polymerase inhibitors. Also used in the determination of cobalt, copper, nickel, and zinc.

Solubility
Soluble in methanol and dimethyl sulfoxide.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 5794-88-7
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007823
InChI Key CUKXRHLWPSBCTI-UHFFFAOYSA-N
Synonym 5-bromoanthranilic acid, benzoic acid, 2-amino-5-bromo, anthranilic acid, 5-bromo, 2-amino-5-bromo-benzoic acid, 5-bromo-2-aminobenzoic acid, 2-amino-5-bromobenzoicacid, 2-amino-5-bromo benzoic acid, 5-bromo anthranilic acid, buttpark 89\07-49, 5-bromo-2-amino-benzoic acid
PubChem CID 79858
IUPAC Name 2-amino-5-bromobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)N

Specifications

Melting Point ∼215°C (decomposition)
Quantity 5 g
Beilstein 639028
Sensitivity Light sensitive
Merck Index 14,1405
Solubility Information Soluble in methanol and dimethyl sulfoxide.
Formula Weight 216.03
Percent Purity 98%
Chemical Name or Material 2-Amino-5-bromobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 227-338-3
RTECSNumber CB2557670
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only