missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromobenzoic acid, 97%

Catalog No. AC305620050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC305620050 5 g
AC305620250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC305620050 Supplier Thermo Scientific Chemicals Supplier No. 305620050

CAS: 5794-88-7 | C7H6BrNO2 | 216.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5794-88-7
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00007823
InChI Key CUKXRHLWPSBCTI-UHFFFAOYSA-N
Synonym 5-bromoanthranilic acid, benzoic acid, 2-amino-5-bromo, anthranilic acid, 5-bromo, 2-amino-5-bromo-benzoic acid, 5-bromo-2-aminobenzoic acid, 2-amino-5-bromobenzoicacid, 2-amino-5-bromo benzoic acid, 5-bromo anthranilic acid, buttpark 89\07-49, 5-bromo-2-amino-benzoic acid
PubChem CID 79858
IUPAC Name 2-amino-5-bromobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 218°C to 220°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H3(Br)CO2H
Quantity 5 g
Merck Index 15, 1412
Formula Weight 216.03
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Amino-5-bromobenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 227-338-3

RUO – Research Use Only